Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human B7-H3 (4Ig)/B7-H3b Protein

Catalog No. 2318B3050
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
2318B3050 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
2318B3050CF Lyophilized from a 0.2 μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 2318B3050 Supplier R&D Systems™ Supplier No. 2318B3050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit anti-CD3-induced proliferation of stimulated human T cells. T lymphocytes cultured for 72 hours with PHA were incubated for an additional 3 days in 96 well plates coated with 500 ng/mL anti-CD3 and/or 10 μg/mL rhB7-H3 (4Ig)/B7-H3b. The presence of rhB7-H3 (4Ig)/B7-H3b at 10 μg/mL inhibited the anti-CD3 response by >30%. Optimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number Q5ZPR3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 80381
Molecular Weight (g/mol) M.W. (Observed): 78-90 kDa, reducing conditions; M.W. (theoretical): 48 kDa
Name B7-H3 (4Ig)/B7-H3b
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human B7-H3 protein Gly27-Thr461, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD276
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein B7-H3/CD276
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.