Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human B7-H4 Protein Fc Chimera

Catalog No. 8870B7050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8870B7050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8870B7050 Supplier R&D Systems™ Supplier No. 8870B7050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to inhibit anti-CD3 antibody induced IL-2 secretion in human T lymphocytes. The ED50 for this effect is 0.1-0.5 μg/mL.

Specifications

Accession Number Q7Z7D3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 79679
Molecular Weight (g/mol) M.W. (theoretical): 52 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human B7-H4 protein Human B7-H4 (Phe29-Ala258) (N-terminus) Accession # Q7Z7D3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1
Common Name B7-H4
Gene Symbol VTCN1
Biological Activity Measured by its ability to inhibit anti-CD3 antibody induced IL-2 secretion in human T lymphocytes. The ED50 for this effect is typically 0.1-0.5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Expression System Human embryonic kidney cell, HEK293-derived Human B7-H4 (Phe29-Ala258) Accession # Q7Z7D3, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.