Learn More
Description
Specifications
Specifications
| Accession Number | P56817.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 23621 |
| Molecular Weight (g/mol) | M.W. (Observed): 68-70 kDa and 65-67 kDa, reducing conditions; M.W. (theoretical): 50 kDa (Pro form) & 47 kDa (Mature form) |
| Name | BACE-1 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived human BACE-1 protein Thr22-Tyr460 (pro) & Glu46-Tyr460 (mature), both with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Pro and Mature forms |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
