Learn More
R&D Systems™ Recombinant Human BAFF/BLyS/TNFSF13B (CHO-expressed) Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 10673 |
| Molecular Weight (g/mol) | 17.0 kDa (monomer) |
| Name | BAFF/BLyS/TNFSF13B |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human BAFF/BLyS/TNFSF13B protein Ala134-Leu285 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | High Molecular Weight Multimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.