Learn More
Description
Specifications
Specifications
| Accession Number | NP_001165886 |
| For Use With (Application) | ELISA, Western Blot |
| Formulation | 20mM Tris-HCl buffer (pH7.5), 100mM NaCl, 10% glycerol |
| Gene ID (Entrez) | 573 |
| Molecular Weight (g/mol) | 25.9kDa |
| Name | Bag-1 Protein |
| Purification Method | Protein |
| Immunogen | MNRSQEVTRD EESTRSEEVT REEMAAAGLT VTVTHSNEKH DLHVTSQQGS SEPVVQDLAQ VVEEVIGVPQ SFQKLIFKGK SLKEMETPLS ALGIQDGCRV MLIGKKNSPQ EEVELKKLKH LEKSVEKIAD QLEELNKELT GIQQGFLPKD LQAEALCKLD RRVKATIEQF MKILEEIDTL ILPENFKDSR LKRKGLVKKV QAFLAECDTV EQNICQETER LQSTNFALAE |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
