Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Band 3 GST (N-Term) Protein
SDP

Catalog No. NB806426 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB806426 25 μg
NB807506 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB806426 Supplier Novus Biologicals™ Supplier No. H00006521Q0125UG
Add to Cart
Add to Cart

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.

Specifications

For Use With (Application) Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 6521
Molecular Weight (g/mol) MolecularWeight-theroretical: 36.74 kDa
Quantity 25 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias AE 1, AE1MGC126619, Anion exchange protein 1, Anion exchanger 1, band 3 anion transport protein, BND3, CD233, CD233 antigen, DI, EMPB3, EPB3MGC126623, erythrocyte membrane protein band 3, erythroid anion exchange protein, FR, Froese blood group, MGC116750, MGC116753, RTA1A, Solute carrier family 4 member 1, solute carrier family 4, anion exchanger, member 1 (erythrocyte membraneprotein band 3, Diego blood group), solute carrier family 4, anion exchanger, number 1, SW, Swann blood group, Waldner blood group, WD, WD1, WR, Wright blood group
Gene Symbol SLC4A1
Research Category Phospho Specific
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Band 3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.