Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BCAT1 Protein

Catalog No. 9536BA020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-36B-A020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 95-36B-A020 Supplier R&D Systems™ Supplier No. 9536BA020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human BCAT1 Protein is derived from E. coli. The Recombinant Human BCAT1 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P54687-1
Characterization Enzyme Activity
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2μm filtered solution in Tris, NaCl, DTT and Glycerol.
Gene ID (Entrez) 586
Molecular Weight (g/mol) 44kDa
Quantity 20 μg
Source E. coli-derived human BCAT1 protein Lys2-Ser386 with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias BCAT(c), BCAT-1, BCATC, BCT1, BCT1PNAS121, branched chain amino-acid transaminase 1, cytosolic, branched chain aminotransferase 1, cytosolic, branched-chain-amino-acid aminotransferase, cytosolic, DKFZp686E12175, EC 2.6.1.42, ECA39, MECA39, placental protein 18, PNAS121, PP18, Protein ECA39
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.