Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Bcl-10 His Protein
SDP

Catalog No. NBP19906410 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19906410 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP19906410 Supplier R&D Systems™ Supplier No. NBP19906410UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-233 of Human Bcl-10 The Recombinant Human Bcl-10 Protein is derived from E. coli. The Recombinant Human Bcl-10 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number O95999
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol, 0.05M NaCl, 1mM DTT
Gene ID (Entrez) 8915
Molecular Weight (g/mol) 28/8kDa
Name Bcl-10 Protein
Purification Method SDS-PAGE
Quantity 10 μg
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Product Line Novus
Purity or Quality Grade >85%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.