Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl and Glycerol. |
| Gene ID (Entrez) | 599 |
| Molecular Weight (g/mol) | 19.5 kDa |
| Name | Bcl-w Minus C-Terminus |
| Quantity | 50 μg |
| Source | E. coli-derived human Bcl-w protein Ala2-Thr172,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Monomer and disulfide-linked homodimer |
| Endotoxin Concentration | <0.01 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
