Learn More
R&D Systems™ Recombinant Human BCMA/TNFRSF17 Fc Alexa Fluor™ 647 Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Accession Number | Q02223.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 608 |
| Molecular Weight (g/mol) | M.W.-observed: 35-45 kDa, reducing conditions, M.W.-theroretical: 32 kDa (monomer) |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human BCMA/TNFRSF17 protein Human BCMA (Met1-Ala54) (N-terminus) Accession # Q02223.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.