Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BDNF Protein
SDP

Catalog No. 248BDB250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
5 μg
50 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
248BDB250 250 μg
248BDB005 5 μg
24-8BD-B010 10 μg
24-8BD-B050 50 μg
248BDB01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 248BDB250 Supplier R&D Systems™ Supplier No. 248BDB250

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human BDNF Protein is derived from Sf 21 (baculovirus). The Recombinant Human BDNF Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P23560
For Use With (Application) Bioactivity
Gene ID (Entrez) 627
Molecular Weight (g/mol) 13-14 kDa, under reducing conditions.
Quantity 250 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived BDNF protein His129-Arg247 Accession &Num; P23560 100% sequence homology with Mouse,Rat,Canine,Equine and all other mammalian proteins examined.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity BDNF
Conjugate Unconjugated
Species Human, Mouse, Rat, Canine, Equine
Recombinant Recombinant
Reconstitution Reconstitute 5 μg vials at 100 μg/mL in water. Reconstitute 10 μg or larger vials at 250 μg/mL in water.
Buffer Lyophilized from a 0.2 μm filtered solution in Sodium Citrate and NaCl with BSA as a carrier protein.
Product Line R&D Systems
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.