Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BDNF Protein, CF

Catalog No. BTBDNF100
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
50 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTBDNF100 100 μg
BTBDNF250 250 μg
BTBDNF050 50 μg
BTBDNF020 20 μg
BTBDNF01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. BTBDNF100 Supplier R&D Systems™ Supplier No. BTBDNF100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using BaF mouse pro-B cells transfected with TrkB.

The ED50 for this effect is 0.100-1.00 ng/mL.

Specifications

Accession Number P23560.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 627
Molecular Weight (g/mol) MolecularWeight-observed: 12 kDa, under reducing conditions., MolecularWeight-theroretical: 13.1 kDa
Quantity 100 μg
Source E. coli-derived human BDNF protein Arg134-Arg247, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin
Conjugate Unconjugated
Reconstitution Reconstitute the 20 μg size at 200 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein BDNF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.