Learn More
Abcam Recombinant Human Beta 2 Adrenergic Receptor Protein, Full Length, Wheat Germ, 2 μg
Description
- Protein Type: Recombinant full length human beta 2 Adrenergic Receptor (1 to 413 aa)
- Expression System: Wheat germ
- Tested Applications: WB
- Form: Liquid
- Storage Buffer: pH 8, containing 0.79% Tris HCl and 0.31% Glutathione
- Purification Technique: Proprietary technique
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Label or Dye | Unconjugated |
| Target Molecule | Beta 2 Adrenergic Receptor (Human) |
| Shipping Condition | Dry Ice |
| Detection Location | In-Blot Detection |
| Buffer | 0.79% Tris-HCl, 0.31% glutathione, pH8.0 |
| Content And Storage | Store at -80°C. |
| Form | Liquid |
| Cross Reactivity | Human |
| Formulation | 0.79% Tris HCl, 0.31% Glutathione, pH 8 |
| Quantity | 2 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.