Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human beta-Catenin His (C-Term) Protein
SDP

Catalog No. NB171820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171820 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171820 Supplier Novus Biologicals™ Supplier No. NBP318198
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. Immobilized Human CD31 at 1 ug/mL (100 ul/well) can bind Human beta-Catenin with a linear range of 0.3-4.9 ug/mL.

Specifications

Concentration LYOPH
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized from a 0.22 um filtered solution of 50mM Tris, 150mM NaCl, pH 8.0
Gene ID (Entrez) 1499
Quantity 100 μg
Storage Requirements Store at -20°C.
Endotoxin Concentration <1.0 EU/μg of the protein by LAL method
Gene Alias beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Gene Symbol CTNNB1
Reconstitution Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water
Research Category Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Purity or Quality Grade >90%, by SDS-PAGE
Protein beta-Catenin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.