Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Betacellulin Protein

Catalog No. BTBTC010
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTBTC010 10 μg
BTBTC01M 1 mg
BTBTC050 50 μg
BTBTC250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. BTBTC010 Supplier R&D Systems™ Supplier No. BTBTC010
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblast cells. Rubin, J.S. et al. (1991) Proc. Natl. Acad. Sci. USA 88:415. The ED50 for this effect is 0.100-1.50 ng/mL.

Specifications

Accession Number P35070.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 685
Molecular Weight (g/mol) MolecularWeight-observed: 12-15 kDa, under reducing conditions., MolecularWeight-theroretical: 9.9 kDa
Quantity 10 μg
Source E. coli-derived human Betacellulin/BTC protein Asp32-Gln118
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias betacellulin, BTC, probetacellulin
Conjugate Unconjugated
Reconstitution Reconstitute at 100-500 μg/mL in PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Betacellulin/BTC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.