Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BID (Caspase-8-cleaved) Protein

Catalog No. 882B8050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
882B8050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 882B8050 Supplier R&D Systems™ Supplier No. 882B8050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to induce cytochrome c release from isolated mouse liver mitochondria using the Rat/Mouse Cytochrome c Quantikine ELISA (Catalog # MCTC0 ) to quantify cytochrome c. The typical EC50 for cytochrome c releasing activity is <50 nM. The EC50 for the desired application should be determined. Uncleaved Recombinant Human BID (Catalog # 846-BD ) is available.

Specifications

Accession Number P55957
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in HEPES-KOH and KCl.
Gene ID (Entrez) 637
Molecular Weight (g/mol) M.W. (Observed): 9 kDa &14 kDa, reducing conditions; M.W. (theoretical): 7 kDa (N-terminal fragment) & 15 kDa (C-terminal fragment)
Name BID Caspase-8-cleaved
Quantity 50 μg
Source E. coli-derived human BID protein Met1-Asp195, protein was purified, cleaved by Caspase-8 and further purified
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Gene Symbol BID
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein BID
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.