Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BMP-2/BMP-6 Heterodimer Protein

Catalog No. 7145BP010
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
7145BP010 Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
7145BP010CF Lyophilized from a 0.2μm filtered solution in HCl.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 7145BP010 Supplier R&D Systems™ Supplier No. 7145BP010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human BMP-2/BMP-6 Heterodimer Protein is derived from E. coli. The Recombinant Human BMP-2/BMP-6 Heterodimer Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Gene ID (Entrez) 650
Molecular Weight (g/mol) 12.8 kDa (BMP-2) & 15 kDa (BMP-6)
Name BMP-2/BMP-6 Heterodimer
Quantity 10 μg
Source E. coli-derived human BMP-2/BMP-6 Heterodimer protein, (N-terminus) Human BMP-2 (Ala284-Arg396),with an N-terminal Met Accession &Num; P12643 Human BMP-6 (Gln382-His513),with an N-terminal Met Accession &Num; P22004 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.