Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BMP-2/BMP-7 Heterodimer Protein

Catalog No. 3229BM010CF
Change view
Click to view available options
Formulation:
Supplied as 0.2 μm filtered solution in 30%(v/v) Acetonitrile and 0.1%(v/v) TFA with 50 μg BSA / 1 μg as a carrier protein.
Supplied as a 0.2 μm filtered solution in Acetonitrile and TFA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
3229BM010CF Supplied as a 0.2 μm filtered solution in Acetonitrile and TFA.
3229BM010 Supplied as 0.2 μm filtered solution in 30%(v/v) Acetonitrile and 0.1%(v/v) TFA with 50 μg BSA / 1 μg as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 3229BM010CF Supplier R&D Systems™ Supplier No. 3229BM010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Nakamura, K. et al. (1999) Exp. Cell Res. 250:351. The ED50 for this effect is 15.0-120 ng/mL.

Specifications

Formulation Supplied as a 0.2 μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 650
Molecular Weight (g/mol) 12.9 kDa (BMP-2), 15.8 kDa (BMP-7)
Name BMP-2/BMP-7 Heterodimer
Quantity 10 μg
Source E. coli-derived human BMP-2/BMP-7 Heterodimer protein, (N-terminus) Human BMP-2 (Ala284 – Arg396) Accession &Num; NP_001191 Human BMP-7 (Ser293 – His431) Accession &Num; NP_001710 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Disulfide-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.