Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BMP-2 GMP Protein, CF
SDP

Catalog No. 355GMP050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-5GM-P050 50 μg
35-5GM-P010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-5GM-P050 Supplier R&D Systems™ Supplier No. 355GMP050
Add to Cart
Add to Cart

Recombinant Protein

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 40-200 ng/mL. The specific activity of recombinant human BMP-2 is >5.0 x 105 units/mg, which is calibrated against the human BMP-2 WHO Standard (NIBSC code: 93/574).

Specifications

Accession Number P12643.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid.
Gene ID (Entrez) 650
Molecular Weight (g/mol) Observed: 15-16, kDa, reducing conditions, Observed: 13 kDa (monomer)
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human BMP-2 protein Gln283-Arg396 Manufactured and tested under current Good Manufacturing Practice (GMP) guidelines.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 12 months when stored at ≤ -20°C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Regulatory Status GMP
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC
Gene Symbol BMP2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining, under reducing conditions
Protein BMP-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.