Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BMP-4 Animal-Free Protein
SDP

Catalog No. AFL314E010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFL314E010 10 μg
AFL314E01M 1 mg
AFL314E250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AFL314E010 Supplier R&D Systems™ Supplier No. AFL314E010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to induce BMP responsive SEAP reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 3-21 ng/mL.

Specifications

Accession Number P12644.1
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 652
Molecular Weight (g/mol) M.W.-observed: 8-9 kDa, under reducing conditions, M.W.-theoretical: 12 kDa
Quantity 10 μg
Source E. coli-derived human BMP-4 protein Lys303-Arg408 Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias BMP2B, BMP-2 B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2 B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Conjugate Unconjugated
Reconstitution Reconstitute at 100-200 μg/mL in 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein BMP-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.