Learn More
R&D Systems™ Recombinant Human BMP-4 Animal-Free Protein
Description
Specifications
Specifications
| Accession Number | P12644.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. |
| Gene ID (Entrez) | 652 |
| Molecular Weight (g/mol) | M.W.-observed: 8-9 kDa, under reducing conditions, M.W.-theoretical: 12 kDa |
| Quantity | 1 mg |
| Source | E. coli-derived human BMP-4 protein Lys303-Arg408 Produced using non-animal reagents in an animal-free laboratory. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | BMP2B, BMP-2 B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2 B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.