Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BMP-4 (E. coli-expressed) Protein

Catalog No. 314BPE050
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-4BP-E050 50 μg
31-4BP-E010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 31-4BP-E050 Supplier R&D Systems™ Supplier No. 314BPE050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells.

Specifications

Accession Number P12644
For Use With (Application) Bioactivity
Gene ID (Entrez) 652
Molecular Weight (g/mol) 12 kDa
Name BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Quantity 50 μg
Source E. coli-derived human BMP-4 protein Lys303-Arg408
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in 4 mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.