Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Measured by its ability to induce BMP responsive SEAP reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 0.70-7.00 ng/mL. Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 3.00-30.0 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
| Gene ID (Entrez) | 652 |
| Molecular Weight (g/mol) | M.W. (Observed): 19-24 kDa, reducing conditions 35-41 kDa, non-reducing conditions |
| Name | BMP-4 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human BMP-4 protein Ser293-Arg408 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
