Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BMP-4 Protein

Catalog No. 314BP050CF
Change view
Click to view available options
Quantity:
10 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
314BP050CF 50 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
314BP010 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
314BP050 50 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
314BP010CF 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
31-4BP-500 500 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
31-4BP-500CF 500 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 314BP050CF Supplier R&D Systems™ Supplier No. 314BP050/CF
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce BMP responsive SEAP reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 0.70-7.00 ng/mL. Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 3.00-30.0 ng/mL.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 652
Molecular Weight (g/mol) M.W. (Observed): 19-24 kDa, reducing conditions 35-41 kDa, non-reducing conditions
Name BMP-4
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human BMP-4 protein Ser293-Arg408
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied.
Structural Form Disulfide-linked homodimer
Gene Alias BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.