Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BMP-7 Biotinylated Protein

Catalog No. BT354B010
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
BT354B010 Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
BT354B010CF Lyophilized from a 0.2μm filtered solution in HCl.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BT354B010 Supplier R&D Systems™ Supplier No. BT354B010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human BMP-7 Biotinylated Protein is derived from CHO. The Recombinant Human BMP-7 Biotinylated Protein has been validated for the following applications: Binding Activity.

Specifications

Accession Number P18075
Characterization Binding Activity
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Gene ID (Entrez) 655
Molecular Weight (g/mol) 16kDa (unlabeled)
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived human BMP-7 protein Ser293-His431
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01EU per 1μg of the protein by the LAL method.
Gene Alias BMP7, BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1, OP-1OP1, Osteogenic protein 1
Product Type Recombinant Protein
Conjugate Biotin
Species Human
Reconstitution Reconstitute at 100μg/mL in 4 mM HCl.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.