Learn More
Specifications
Specifications
| Accession Number | P18075, P18075, P18075 |
| Concentration | LYOPH |
| For Use With (Application) | Western Blot, ELISA, PAGE |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 30 % acetonitrile, 0.1 % TFA. |
| Gene ID (Entrez) | 655 |
| Molecular Weight (g/mol) | 15.7 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of BMP-7 as determined by LAL method. |
| Gene Alias | BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
