Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BST-2/Tetherin Protein
SDP

Catalog No. 9939BS050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
99-39B-S050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 99-39B-S050 Supplier R&D Systems™ Supplier No. 9939BS050
Add to Cart
Add to Cart
This item is not returnable. View return policy

BST-2, also known as tetherin or CD317, is a type II transmembrane protein that is constitutively expressed in several cell types including T cells, B cells, monocytes and macrophages, as well as on several cancer cell lines including the B cell lineage in multiple myeloma

  • Recombinant Protein
  • Carrier Free (CF)
  • The carrier free version does not contain Bovine Serum Albumin (BSA)

Specifications

Accession Number Q10589-1
For Use With (Application) Bioactivity
Gene ID (Entrez) 684
Molecular Weight (g/mol) 14 kDa
Purification Method >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source E. coli-derived human BST-2/Tetherin protein Asn49-Ser161, with an N-terminal Met and C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water
Buffer Lyophilized from a 0.2 μm filtered solution in MES and NaCl with Trehalose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.