Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BTLA Fc Chimera (aa 25-150) Protein

Catalog No. 8385BT050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-85B-T050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-85B-T050 Supplier R&D Systems™ Supplier No. 8385BT050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Mouse HVEM/TNFRSF14 Fc Chimera (Catalog # 2516-HV ) is immobilized at 0.5 μg/mL (100 μL/well), the concentration of Recombinant Human BTLA Fc Chimera that produces 50% of the optimal binding response is approximately 0.25-1.25 μg/mL.

Specifications

Accession Number AAP44003
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 151888
Molecular Weight (g/mol) M.W. (Observed): 53-65 kDa, reducing conditions; M.W. (theoretical): 41 kDa
Name BTLA Fc Chimera
Preparation Method Reconstitute at 500μg/mL in PBS.
Quality Control Testing SDS-PAGE: 53-65kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human BTLA protein Human BTLA Ile25-Ser150 (N-terminus) Accession # AAP44003 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ile25
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol BTLA
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >85%, by SDS-PAGE with silver staining.
Protein BTLA/CD272
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.