Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BTN2A1/Butyrophilin 2A1 Fc Avi Protein

Catalog No. AVI10909050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AVI10909050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AVI10909050 Supplier R&D Systems™ Supplier No. AVI10909050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Biotinylated

Measured by its binding ability in a functional ELISA. When Recombinant Human CD277/BTN3A1 Fc Chimera Protein (Catalog # 8539-BT ) is immobilized at 10.0 μg/mL, 100 μL/well, the concentration of Biotinylated Recombinant Human BTN2A1 Fc Chimera Avi-tag (Catalog #AVI10909) that produces 50% of the binding response is 1.00-6.00 μg/mL.

Specifications

Accession Number Q7KYR7.3
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 11120
Molecular Weight (g/mol) M.W.-observed: 65-80 kDa, under reducing conditions., M.W.-theroretical: 53 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human BTN2A1 protein Human BTN2A1 (Gln29-Ala248) (N-terminus) Accession # Q7KYR7.3 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias BK14H9.1, BT2.1, BTF1, BTF1BT2.1 butyrophilin BTF1, butyrophilin subfamily 2 member A1, butyrophilin, subfamily 2, member A1, DJ3E1.1, FLJ36567
Conjugate Biotin
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein BTN2A1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.