Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BTN2A2/Butyrophilin 2A2 Fc Protein
SDP

Catalog No. 8918BT050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8918BT050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8918BT050 Supplier R&D Systems™ Supplier No. 8918BT050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Learn More About B7-related Butryophilins as Immune Modulators View Webinar

Measured by its ability to inhibit anti-CD3 antibody induced IL-2 secretion in human T lymphocytes. The ED50 for this effect is 0.1-0.5 μg/mL.

Specifications

Accession Number Q8WVV5
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 10385
Molecular Weight (g/mol) M.W. (theoretical): 50 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human BTN2A2/Butyrophilin 2A2 protein Human BTN2A2 (Gln33-Val237) (N-terminus) Accession # Q8WVV5 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias BTF2, BTF2BT2.2butyrophilin 2, BTN2, Butyrophilin 2, butyrophilin subfamily 2 member A2, butyrophilin, subfamily 2, member A2, FLJ41908
Common Name BTN2A2/Butyrophilin 2
Gene Symbol BTN2A2
Biological Activity Measured by its ability to inhibit anti-CD3 antibody induced IL-2 secretion in human T lymphocytes. The ED50 for this effect is typically 0.1-0.5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Expression System Human embryonic kidney cell, HEK293-derived Human BTN2A2 (Gln33-Val237) Accession # Q8WVV5, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein BTN2A2/Butyrophilin 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.