Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BTNL8 Fc Chimera Biotinylated Protein, CF

Catalog No. BT9359050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT9359050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT9359050 Supplier R&D Systems™ Supplier No. BT9359050
This item is not returnable. View return policy

Learn More About B7-related Butryophilins as Immune Modulators View Webinar

Measured by its ability to enhance anti-CD3-induced IFN-gamma secretion of mouse CD3+ T cells. The ED50 for this effect is 0.5-2.5 μg/mL.

Specifications

Accession Number Q6UX41-1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 79908
Molecular Weight (g/mol) M.W. (Observed): 56-63 kDa and 100 - 160 kDa, reducing conditions; M.W. (theoretical): 52 kDa (unlabeled)
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human BTNL8 protein Human BTNL8 (Gln18-Lys238) (N-terminus) Accession # Q6U x 41-1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Biotin
Cross Reactivity BTNL8
Species Human
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein BTNL8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.