Learn More
R&D Systems™ Recombinant Human c-Abl His-tag Protein
Description
Specifications
Specifications
| Accession Number | P00519.4 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, TCEP and Glycerol. |
| Gene ID (Entrez) | 25 |
| Molecular Weight (g/mol) | M.W.-observed: 33-35 kDa, under reducing conditions, M.W.-theroretical: 34 kDa |
| Quantity | 20 μg |
| Source | E. coli-derived human c-Abl protein Gly227-Gln513, with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Abelson murine leukemia viral oncogene homolog 1, ABL, ABL1, bcr/abl, BCR-ABL1, cAbl, c-ABL, c-abl oncogene 1, non-receptor tyrosine kinase, c-abl oncogene 1, receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.2, JTK7, JTK7bcr/abl, p150bcr/c-abl oncogene protein, Proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, tyrosine-protein kinase ABL1, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.