Learn More
Abcam Recombinant Human c-Myc Protein, Full Length, 5 μg
Description
- Protein Type: Recombinant
- Species: Human
- Expression System: Escherichia coli
- Purity: >75%
- Applications: SDS-PAGE, WB
- Formulation: Liquid. pH 7.9. Constituents: EDTA, Potassium chloride, Tris HCl, 20% Glycerol, and 0.0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Liquid |
| Detection Location | In-Blot Detection, In-Gel Detection |
| Label or Dye | Unconjugated |
| Buffer | Tris HCl, EDTA, potassium chloride, 20% glycerol, 0.0154% (R*,R*)-1,4-dimercaptobutan-2,3-diol, pH7.9 |
| Content And Storage | Store at -80°C. |
| Shipping Condition | Dry Ice |
| Target Molecule | c-Myc (Human) |
| Format | Vial |
| Reactivity | Human |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.