Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human C-Reactive Protein/CRP Protein

Catalog No. 1707CR200
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Tris-HCl, NaCl and CaCl 2 .
Lyophilized from a 0.2 μm filtered solution in Tris-HCl, NaCl and CaCl 2 with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
1707CR200 Lyophilized from a 0.2 μm filtered solution in Tris-HCl, NaCl and CaCl 2 with BSA as a carrier protein.
1707CR200CF Lyophilized from a 0.2 μm filtered solution in Tris-HCl, NaCl and CaCl 2 .
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 1707CR200 Supplier R&D Systems™ Supplier No. 1707CR200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Human C-Reactive Protein/CRP Protein is derived from NS0. The Recombinant Human C-Reactive Protein/CRP Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCl, NaCl and CaCl 2 with BSA as a carrier protein.
Gene ID (Entrez) 1401
Molecular Weight (g/mol) 23 kDa (monomer)
Name C-Reactive Protein/CRP
Quantity 200 μg
Source Mouse myeloma cell line,NS0-derived human C-Reactive Protein/CRP protein Phe17-Pro224
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Homopentamer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.