Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human C1qTNF11/C1qL4 Protein
SDP

Catalog No. 9758TN050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
97-58T-N050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 97-58T-N050 Supplier R&D Systems™ Supplier No. 9758TN050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human C1qTNF11/C1qL4 Protein is derived from E. coli. The Recombinant Human C1qTNF11/C1qL4 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q86Z23
For Use With (Application) Bioactivity
Gene ID (Entrez) 338761
Molecular Weight (g/mol) 16 kDa
Quantity 50 μg
Source E. coli-derived human C1qL4 protein Gly105-Asp238,with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity C1QL4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in water.
Buffer Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and TCEP with Trehalose.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.