Learn More
Specifications
Specifications
| Concentration | 0.25 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20 mM Tris-HCl buffer (pH8.5), 30% glycerol, 0.2 M NaCl |
| Gene ID (Entrez) | 114902 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 26.4 kDa |
| Quantity | 50 μg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Endotoxin Concentration | Less Than 1.0 EU / 1 μg of protein (determined by LAL method) |
| Gene Alias | C1q and tumor necrosis factor related protein 5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3 |
| Gene Symbol | C1QTNF5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
