Learn More
Abcam Recombinant Human C3 Protein, Wheat Germ, 10 μg
Description
- Protein Type: Recombinant Human C3 protein
- Expression System: Wheat germ
- Tested Applications: WB, ELISA
- Fragment Range: 1534 to 1644 aa
- Formulation: Liquid, pH 8
- Storage Buffer Constituents: 0.79% Tris HCl, 0.31% Glutathione
- Purification Technique: Proprietary technique
- Storage Conditions: Store at -80°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | C3 |
| Label or Dye | Unconjugated |
| Shipping Condition | Dry Ice |
| Buffer | 0.79% Tris-HCl, 0.31% Glutathione, pH8 |
| Content And Storage | Store at -80°C. |
| Form | Liquid |
| Formulation | 0.79% Tris HCl, 0.31% Glutathione, pH 8 |
| Quantity | 10 μg |
| For Use With (Application) | ELISA, Western Blot |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.