Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 28316 |
| Molecular Weight (g/mol) | 65.7 kDa (pro) & 62.8 kDa (mature) |
| Name | Cadherin-20 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Cadherin-20 protein Asp35-Arg619 (pro) & Ser60-Arg619 (mature),both with a C-terminal 6-His tag The two forms are approximately in equal ratio. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
