Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Calcium-sensing R/CaSR Protein

Catalog No. 9608CS050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9608CS050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9608CS050 Supplier R&D Systems™ Supplier No. 9608CS050
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Human Calcium-sensing R/CaSR Protein CF is a plasma membrane G-protein-coupled receptor that senses changes in the extracellular concentration of calcium ions and plays a key role in maintaining calcium homeostasis.

The Recombinant Human Calcium-sensing R/CaSR Protein is derived from HEK293.

Specifications

Accession Number P41180-1
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 846
Molecular Weight (g/mol) 66 kDa
Name calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2A, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human Calcium-sensing R/CaSR protein Tyr20-Lys601,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.