Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Calmodulin 2 Protein
SDP

Catalog No. NB287363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287363 0.5 mg
NB287360 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB287363 Supplier Novus Biologicals™ Supplier No. NBP3215030.5MG
Only null left
Add to Cart
Add to Cart

Specifications

Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5)
Gene ID (Entrez) 805
Molecular Weight (g/mol) 16 kDa
Quantity 0.5 mg
Storage Requirements Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Gene Alias CALM, CALM1, CALM3, CALML2, calmodulin, calmodulin 2 (phosphorylase kinase, delta), CAM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMII, CAMIII, FLJ99410, LP7057 protein, PHKD, PHKD2, phosphorylase kinase delta
Gene Symbol CALM2
Research Category Cancer
Purity or Quality Grade >90%, by SDS Page
Protein Calmodulin 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.