Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Carboxylesterase 2/CES2 Protein
SDP

Catalog No. 5657CE010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5657CE010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5657CE010 Supplier R&D Systems™ Supplier No. 5657CE010

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to hydrolyze p-nitrophenylacetate. The specific activity is >30,000 pmol/min/μg, measured under the described conditions.

Specifications

Accession Number O00748
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Sodium Acetate, NaCl and Glycerol.
Gene ID (Entrez) 8824
Molecular Weight (g/mol) M.W. (Observed): 63 kDa, reducing conditions; M.W. (theoretical): 60 kDa
Name Carboxylesterase 2/CES2
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Carboxylesterase 2/CES2 protein Gln27-Leu559, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Structural Form Monomer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol CES2
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Carboxylesterase 2/CES2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.