Learn More
Description
Specifications
Specifications
| Accession Number | Q96KN2 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 84735 |
| Molecular Weight (g/mol) | M.W. (Observed): 65 kDa, reducing conditions; M.W. (theoretical): 55 kDa |
| Name | Carnosine Dipeptidase 1/CNDP1 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived human Carnosine Dipeptidase 1/CNDP1 protein Pro28-His507, with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
