Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Caspase-3 Protein
SDP

Catalog No. 707C3010CF Shop All Novus Biologicals Products
Change view
Click to view available options
Formulation:
Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
707C3010CF Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 707C3010CF Supplier R&D Systems™ Supplier No. 707C3010/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Caspase-3 Protein is derived from E. coli. The Recombinant Human Caspase-3 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
Gene ID (Entrez) 836
Molecular Weight (g/mol) 17 kDa (subunit 1), 11 kDa (subunit 2)
Name Caspase-3
Quantity 10 μg
Source E. coli-derived human Caspase-3 protein Ser29-Asp175 (subunit 1) & Ala183-His277 (Asp190Glu) (subunit 2)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.