Learn More
Description
Specifications
Specifications
| Accession Number | P04040 |
| Concentration | 1mg/mL |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 10% glycerol |
| Gene ID (Entrez) | 847 |
| Quantity | 100μg |
| Storage Requirements | Store at 4° short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Recombinant |
| Purity or Quality Grade | >90%, by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
