Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Cathepsin E Protein

Catalog No. 1294AS010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1294AS010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1294AS010 Supplier R&D Systems™ Supplier No. 1294AS010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2 (Catalog # ES001 ). The specific activity is >1,500 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number NP_001901
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in MES and NaCl.
Gene ID (Entrez) 1510
Molecular Weight (g/mol) M.W. (Observed): 48 kDa, reducing conditions; M.W. (theoretical): 42 kDa
Name Cathepsin E
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Cathepsin E protein Gln18-Pro396, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Pro and mature forms
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol CTSE
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 25 mM MES, 150 mM NaCl, pH 6.5.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Cathepsin E
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.