Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in MES, NaCl and Glycerol. |
| Gene ID (Entrez) | 1520 |
| Molecular Weight (g/mol) | 39 kDa (Pro) and 28 kDa (mature) |
| Name | Cathepsin S |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Cathepsin S protein Gln17-Ile331 (pro) & Ser109-Ile331 (mature),both with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Pro and mature forms |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
