Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Sodium Acetate and NaCl. |
| Gene ID (Entrez) | 1522 |
| Molecular Weight (g/mol) | 33 kDa |
| Name | Cathepsin X/Z/P |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Cathepsin X/Z/P protein Ala21-Val303 (Gly23Val & Ser48Thr),with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Pro form |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
