Learn More
R&D Systems™ Recombinant Human CCL26/Eotaxin-3 (aa 24-94) Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
| Gene ID (Entrez) | 10344 |
| Molecular Weight (g/mol) | 8.4 kDa |
| Name | CCL26/Eotaxin-3 aa 24-94 |
| Quantity | 25 μg |
| Source | E. coli-derived human CCL26/Eotaxin-3 protein Thr24-Leu94 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | CC chemokine IMAC, C-C motif chemokine 26, chemokine (C-C motif) ligand 26, chemokine N1, Eotaxin3, Eotaxin-3, IMAC, Macrophage inflammatory protein 4-alpha, MGC126714, MIP-4a, MIP-4alpha, SCYA26, SCYA26MIP-4-alpha, small inducible cytokine A26, small inducible cytokine subfamily A (Cys-Cys), member 26, Small-inducible cytokine A26, TSC-1Thymic stroma chemokine-1 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.