Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CCT6B GST (N-Term) Protein
SDP

Catalog No. NB812320 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB812320 10 μg
NB812236 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB812320 Supplier R&D Systems™ Supplier No. H00010693P0110UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 10693
Molecular Weight (g/mol) 84.04 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias Cctz2, CCTZ-2, CCT-zeta-2, CCT-zeta-like, chaperonin containing TCP1, subunit 6B (zeta 2), T-complex protein 1 subunit zeta-2, T-complex protein 1, zeta-2 subunit, TCP-1-zeta-2, TCP-1-zeta-like, Testis-specific protein TSA303, Testis-specific Tcp20, TSA303
Gene Symbol CCT6B
Research Category Signal Transduction
Product Line Novus
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein CCT6B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.