Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD155/PVR Fc Chimera Avi-tag Protein

Catalog No. AVI9174050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AVI9174050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AVI9174050 Supplier R&D Systems™ Supplier No. AVI9174050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Biotinylated

Measured by its binding ability in a functional ELISA. When Recombinant Human CD96v2 Fc Chimera (Catalog # 9556-CD ) is immobilized at 1 μg/mL (100 μL/well), Biotinylated Recombinant Human CD155/PVR Fc Chimera Avi-tag (Catalog # AVI9174) binds with an ED50 of 10-60 ng/mL.

Specifications

Accession Number NP_006496.4
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 5817
Molecular Weight (g/mol) M.W. (Observed): 86-98 kDa, reducing conditions; M.W. (theoretical): 63 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human CD155/PVR protein Human CD155/PVR (Asp28-Asn343) (N-terminus) Accession # NP_006496.4 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity CD155/PVR
Conjugate Biotin
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD155/PVR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.